Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Irwin Barricade Tape, 'CAUTION' Black/Yellow, 2 mil, 1 Roll

Catalog No. 501909751
Encompass

This barricade tape is designed for temporary hazard marking, featuring a black 'CAUTION' legend on a yellow background. Constructed from durable 2 mil polyethylene, it provides clear visual warning and measures 3 in width and 300 ft in length.

  • Tape type: Barricade
  • Legend: CAUTION
  • Material: Polyethylene
  • Thickness: 2 mil
  • Roll length: 300 ft
  • Roll width: 3 in
  • Color: Yellow, black
  • Language: English

Catalog No. 50-190-9751 Supplier MSC Supplier No. 77274736
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This barricade tape is designed for temporary hazard marking, featuring a black 'CAUTION' legend on a yellow background. Constructed from durable 2 mil polyethylene, it provides clear visual warning and measures 3 in width and 300 ft in length.

  • Tape type: Barricade
  • Legend: CAUTION
  • Material: Polyethylene
  • Thickness: 2 mil
  • Roll length: 300 ft
  • Roll width: 3 in
  • Color: Yellow, black
  • Language: English

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.