Learn More
Medchemexpress LLC HY-119373 5mg Medchemexpress, Cav 2.2 blocker 1 CAS:1567335-29-8 Purity:>98%
SDP
Catalog No. 501932537
Encompass_Preferred
Medchemexpress, HY-119373 5mg Cav 2.2 blocker 1 CAS:1567335-29-8 Cav 2.2 blocker 1 (compound 9) is a N-type calcium channel (Cav 2.2) blocker for the treatment of pain, with an IC 50 of 1 nM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2537 Supplier Medchemexpress LLC Supplier No. HY1193735MG
Medchemexpress, HY-119373 5mg Cav 2.2 blocker 1 CAS:1567335-29-8 Cav 2.2 blocker 1 (compound 9) is a N-type calcium channel (Cav 2.2) blocker for the treatment of pain, with an IC 50 of 1 nM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.