Learn More
Aviva Systems Biology Corp CAV1 Antibody - N-terminal region (AVARP09019_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP09019_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CAV1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human CAV1. Alternate names: CAV, MSTP085, VIP21, CGL3, BSCL3ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-62572