Learn More
Aviva Systems Biology Corp CAV3 Antibody - N-terminal region (AVARP09021_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP09021_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CAV3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CAV3. Alternate names: LQT9, VIP21, LGMD1C, VIP-21ApplicationIHC, WB. Species reactivity: Cow, Human, Mouse, Pig, Rat
Catalog No. 50-014-62574