Learn More
Enzo Life Sciences Caveolin 1 scaffolding domain peptide (1mg)
SDP
Supplier:  Enzo Life Sciences ALX153064M001
Encompass_Preferred
Cell permeable peptide that blocks eNOS (NOS III) activity and cellular nitric oxide (NO) release in vitro and reduces inflammation and tumorigenesis in vivo. Caveolin-1 interacts with several lipid-modified signalling ligands, such as EGFR, eNOS, G-protein α-subunits, PKCα, H-Ras, and Src, via the C1-SD82-101 sequence. Alternative name: Cavtratin, AP-Cav, Pen-C1-SD. Formula: C228H335N61O49S. MW: 4746.6. Purity: ≥97% (HPLC). Appearance: White lyophilized powder. Solubility: Soluble in DMSO or acetonitrile:water (50:50). Long Term Storage: -20°C. Use/Stability: Stock solutions are stable for up to 3 months when stored at -20°C. Handling: Protect from light. Hygroscopic.
Catalog No. 50-200-7789