Learn More
Enzo Life Sciences Caveolin 1 scaffolding domain peptide (5mg)
SDP
Catalog No. 502007790
Encompass_Preferred
Cell permeable eNOS blocker. Cell permeable peptide that blocks eNOS (NOS III) activity and cellular nitric oxide (NO) release in vitro and reduces inflammation and tumorigenesis in vivo. Caveolin-1 interacts with several lipid-modified signalling ligands, such as EGFR, eNOS, G-protein α-subunits, PKCα, H-Ras, and Src, via the C1-SD82-101 sequence. Alternative name: Cavtratin, AP-Cav, Pen-C1-SD. Formula: C228H335N61O49S. MW: 4746.6. Purity: ≥97% (HPLC). Appearance: White lyophilized powder. Solubility: Soluble in DMSO or acetonitrile:water (50:50). Long Term Storage: -20°C. Use/Stability: Stock solutions are stable for up to 3 months when stored at -20°C. Handling: Protect from light. Hygroscopic.
Catalog No. 50-200-7790 Supplier Enzo Life Sciences Supplier No. ALX153064M005
Add to Cart
Add to Cart
Cell permeable eNOS blocker. Cell permeable peptide that blocks eNOS (NOS III) activity and cellular nitric oxide (NO) release in vitro and reduces inflammation and tumorigenesis in vivo. Caveolin-1 interacts with several lipid-modified signalling ligands, such as EGFR, eNOS, G-protein α-subunits, PKCα, H-Ras, and Src, via the C1-SD82-101 sequence. Alternative name: Cavtratin, AP-Cav, Pen-C1-SD. Formula: C228H335N61O49S. MW: 4746.6. Purity: ≥97% (HPLC). Appearance: White lyophilized powder. Solubility: Soluble in DMSO or acetonitrile:water (50:50). Long Term Storage: -20°C. Use/Stability: Stock solutions are stable for up to 3 months when stored at -20°C. Handling: Protect from light. Hygroscopic.