Learn More
Enzo Life Sciences Caveolin-3 polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ALX210241C100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to aa 1-18 (M1MTEEHTDLEARIIKDIH18C) of rat caveolin-3. UniProt ID: P51638. Species reactivity: Human, Mouse, Rat. Creossreactivity: Does not cross-react with caveolin-1 or -2. Applications: ICC, IF, IHC, IP, WB. Recommended Dilutions: Immunohistochemistry (1:100-200) Immunofluorescence (1:20) Western Blot (1µg/ml) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Epitope affinity-purified. Formulation: Liquid. In PBS containing 1mg/ml BSA and 0.05% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-8098