Learn More
Cayman Chemical CaveolIn13 Blockng PepID 200ug
SDP
Catalog No. 502978764
Encompass_Preferred
An inhibitor of menin-MLL fusion protein interactions (IC50 446 nM) binds to menin (Kd 158 nM) and inhibits the menin-MLL-AF9 interaction in HEK293 cells inhibits growth of mouse BMCs expressing the Hoxa9 transformation-dependent MLL-AF9 (GI50 5 UM) inhibits the growth of MV4 KOPN-8 ML-2 and MonoMac6 MLL cells expressing MLL fusion proteins (GI50s 9 5 7 2 8 7 and 18 M respectively)
Catalog No. 50-297-8764 Supplier Cayman Chemical Supplier No. 300830200 UG
An inhibitor of menin-MLL fusion protein interactions (IC50 446 nM) binds to menin (Kd 158 nM) and inhibits the menin-MLL-AF9 interaction in HEK293 cells inhibits growth of mouse BMCs expressing the Hoxa9 transformation-dependent MLL-AF9 (GI50 5 UM) inhibits the growth of MV4 KOPN-8 ML-2 and MonoMac6 MLL cells expressing MLL fusion proteins (GI50s 9 5 7 2 8 7 and 18 M respectively)