Learn More
Cayman Chemical CaveolIn1CTRm RabbIt Mo 100uL
SDP
Catalog No. 502946367
Encompass_Preferred
An analytical reference standard that is structurally similar to known synthetic cannabinoids a potential metabolite of 5-fluoro MPP-PICA based on the published metabolism of 5-fluoro AMB-PICA intended for research and forensic applications
Catalog No. 50-294-6367 Supplier Cayman Chemical Supplier No. 32263100 UL
An analytical reference standard that is structurally similar to known synthetic cannabinoids a potential metabolite of 5-fluoro MPP-PICA based on the published metabolism of 5-fluoro AMB-PICA intended for research and forensic applications