Learn More
Newcomer Supply CaviWipes 1 Disinfectant, 160 wipes/canister, 12 canisters/case, Works in 1 minute!
SDP
Catalog No. 501959335
Encompass
CaviWipes1 Surface Disinfection Towelettes are a multi-purpose disinfectant/decontaminant wipe that can be used on hard, non-porous surfaces.  The improvements found in the latest formulation are effective in killing of bacteria, viruses and fungi in 1 minute including TB, MDR A. baumannii, HIV-1, HCV and many more. When used as directed, the fragrance free CaviWipes1 Surface Disinfection Towelettes will effectively clean and disinfect surfaces and can help reduce the risk of cross contamination.
This item is not returnable. View return policy
Catalog No. 501959335 Supplier Newcomer Supply Supplier No. ACP135100
CaviWipes1 Surface Disinfection Towelettes are a multi-purpose disinfectant/decontaminant wipe that can be used on hard, non-porous surfaces.  The improvements found in the latest formulation are effective in killing of bacteria, viruses and fungi in 1 minute including TB, MDR A. baumannii, HIV-1, HCV and many more. When used as directed, the fragrance free CaviWipes1 Surface Disinfection Towelettes will effectively clean and disinfect surfaces and can help reduce the risk of cross contamination.