Learn More
Newcomer Supply CaviWipes Disinfectant, 160 wipes/canister
SDP
Catalog No. NC0779310
Encompass
Pre-saturated with CaviCide, CaviWipes Surface Disinfection Towelettes are a cleaner and disinfectant in one.  CaviWipes Surface Disinfection Towelettes are durable, non-woven, non-abrasive towelettes that offer quick, easy-to-use, time-saving convenience.  They are recommended for use on hard non-porous surfaces and fixtures.  CaviWipes are effective against Staphylococcus aureus with reduced susceptibility to vancomycin and influenza A2 Virus and Kills TB and MRSA in 3 minutes and HIV-1 and HBV and HCV in 2 minutes. CaviWipes Surface Disinfection Towelettes should not be disposed of in the toilet.
This item is not returnable. View return policy
Catalog No. NC0779310 Supplier Newcomer Supply Supplier No. ACP131100
Pre-saturated with CaviCide, CaviWipes Surface Disinfection Towelettes are a cleaner and disinfectant in one.  CaviWipes Surface Disinfection Towelettes are durable, non-woven, non-abrasive towelettes that offer quick, easy-to-use, time-saving convenience.  They are recommended for use on hard non-porous surfaces and fixtures.  CaviWipes are effective against Staphylococcus aureus with reduced susceptibility to vancomycin and influenza A2 Virus and Kills TB and MRSA in 3 minutes and HIV-1 and HBV and HCV in 2 minutes. CaviWipes Surface Disinfection Towelettes should not be disposed of in the toilet.