Learn More
Cayman Chemical Helicid, Purity: ≥95%, CAS: 80154-34-3, 10 g
SDP
Catalog No. 502950309
Encompass_Preferred
A β-allopyranoside with diverse biological activities; reverses decreases in serum CORT levels and the expression of rat hippocampal 5-HT1A receptors in a model of chronic unpredictable mild stress-induced depression at 32 mg/kg per day; has anti-depressant-like effects and improves spatial memory in rat models of chronic mild stress-induced depression; increases the mechanical pain threshold and increases NREM sleep time in a manner dependent on the time-of-day of administration in a mouse model of partial sciatic nerve ligation
Catalog No. 50-295-0309 Supplier Cayman Chemical Supplier No. 3341410 G
A β-allopyranoside with diverse biological activities; reverses decreases in serum CORT levels and the expression of rat hippocampal 5-HT1A receptors in a model of chronic unpredictable mild stress-induced depression at 32 mg/kg per day; has anti-depressant-like effects and improves spatial memory in rat models of chronic mild stress-induced depression; increases the mechanical pain threshold and increases NREM sleep time in a manner dependent on the time-of-day of administration in a mouse model of partial sciatic nerve ligation