Learn More
Medchemexpress LLC CAY 10465 50mg | 688348-33-6 | 98.3% | 317.13 | 50 MG
SDP
Catalog No. 5000436297
Encompass_Preferred
CAY 10465 is a selective and high-affinity AhR agonist, with a Ki of 0.2 nM, and shows no effect on estrogen receptor (Ki >100000 nM).
  • Is a selective and high-affinity AhR receptor agonist, with a Ki of 0.2 nM, and shows no effect on estrogen receptor (Ki >100000 nM).
  • Reduces apolipoprotein A-I (apo A-I) levels in HepG2 cells, and inhibits the synthesis of the protein (0.01 nM-100 μM).
Catalog No. 50-004-36297 Supplier Medchemexpress LLC Supplier No. HY11262750MG
CAY 10465 is a selective and high-affinity AhR agonist, with a Ki of 0.2 nM, and shows no effect on estrogen receptor (Ki >100000 nM).
  • Is a selective and high-affinity AhR receptor agonist, with a Ki of 0.2 nM, and shows no effect on estrogen receptor (Ki >100000 nM).
  • Reduces apolipoprotein A-I (apo A-I) levels in HepG2 cells, and inhibits the synthesis of the protein (0.01 nM-100 μM).