Learn More
Medchemexpress LLC HY-112627 5mg Medchemexpress, CAY 10465 CAS:688348-33-6 Purity:>98%
SDP
Catalog No. 501969122
Encompass_Preferred
Medchemexpress, HY-112627 5mg CAY 10465 CAS:688348-33-6 CAY 10465 is a selective and high-affinity AhR agonist, with a Ki of 0.2 nM, and shows no effect on estrogen receptor (Ki >100000 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9122 Supplier Medchemexpress LLC Supplier No. HY1126275MG
Medchemexpress, HY-112627 5mg CAY 10465 CAS:688348-33-6 CAY 10465 is a selective and high-affinity AhR agonist, with a Ki of 0.2 nM, and shows no effect on estrogen receptor (Ki >100000 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.