Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CAY 10465 | 688348-33-6 | 98.3% | 317.13 | 100 MG
SDP

Catalog No. 5000437567
Encompass_Preferred

CAY 10465 is a selective and high-affinity Aryl Hydrocarbon Receptor (AhR) agonist, with a Ki of 0.2 nM. It demonstrates no effect on the estrogen receptor (Ki >100000 nM). In vitro, CAY 10465 (at concentrations from 0.01 nM to 100 μM) has been shown to reduce apolipoprotein A-I (apo A-I) levels in HepG2 cells and inhibit the synthesis of this protein.

  • Selective AhR agonist: High affinity for AhR with a Ki of 0.2 nM.
  • No estrogen receptor effect: Shows no activity on the estrogen receptor (Ki >100000 nM).
  • Inhibits apolipoprotein A-I: Reduces apo A-I levels and inhibits its synthesis in HepG2 cells.
  • Appearance: Solid, white to off-white.
  • Storage:
    Powder: -20°C for 3 years, 4°C for 2 years.
    In solvent: -80°C for 2 years, -20°C for 1 year.

Catalog No. 50-004-37567 Supplier Medchemexpress LLC Supplier No. HY112627100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAY 10465 is a selective and high-affinity Aryl Hydrocarbon Receptor (AhR) agonist, with a Ki of 0.2 nM. It demonstrates no effect on the estrogen receptor (Ki >100000 nM). In vitro, CAY 10465 (at concentrations from 0.01 nM to 100 μM) has been shown to reduce apolipoprotein A-I (apo A-I) levels in HepG2 cells and inhibit the synthesis of this protein.

  • Selective AhR agonist: High affinity for AhR with a Ki of 0.2 nM.
  • No estrogen receptor effect: Shows no activity on the estrogen receptor (Ki >100000 nM).
  • Inhibits apolipoprotein A-I: Reduces apo A-I levels and inhibits its synthesis in HepG2 cells.
  • Appearance: Solid, white to off-white.
  • Storage:
    Powder: -20°C for 3 years, 4°C for 2 years.
    In solvent: -80°C for 2 years, -20°C for 1 year.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.