Learn More
Cayman Chemical CAY10434, Purity: ≥98%, CAS: 769917-29-5, 10 mg
SDP
Catalog No. 502891127
Encompass_Preferred
A selective inhibitor of the 20-HETE synthase CYP4A11 (IC50 = 8.8 nM human renal microsomes)
Catalog No. 50-289-1127 Supplier Cayman Chemical Supplier No. 1000506710 MG
A selective inhibitor of the 20-HETE synthase CYP4A11 (IC50 = 8.8 nM human renal microsomes)