Learn More
Cayman Chemical CAY10443, Purity: ≥98%, CAS: 582314-48-5, 50 mg
SDP
Catalog No. 502890735
Encompass_Preferred
A pro-apoptotic activator of the apoptosome; activates caspase-3 (EC50 = 5 µM) in a cell-free assay
Catalog No. 50-289-0735 Supplier Cayman Chemical Supplier No. 1000417750 MG
A pro-apoptotic activator of the apoptosome; activates caspase-3 (EC50 = 5 µM) in a cell-free assay