Learn More
Cayman Chemical CAY10443, Purity: ≥98%, CAS: 582314-48-5, 10 mg
SDP
Catalog No. 502888735
Encompass_Preferred
A pro-apoptotic activator of the apoptosome; activates caspase-3 (EC50 = 5 µM) in a cell-free assay
Catalog No. 50-288-8735 Supplier Cayman Chemical Supplier No. 1000417710 MG
A pro-apoptotic activator of the apoptosome; activates caspase-3 (EC50 = 5 µM) in a cell-free assay