Learn More
Cayman Chemical CAY10448 Fatty Acids and Deriv
SDP
Catalog No. 502890873
Encompass_Preferred
An iodinated nonivamide; a TRPV1 antagonist (IC50 = ~10 nM)
Catalog No. 50-289-0873 Supplier Cayman Chemical Supplier No. 100056331 MG
An iodinated nonivamide; a TRPV1 antagonist (IC50 = ~10 nM)