Learn More
Cayman Chemical CAY10448, Purity: ≥98%, CAS: 1177195-52-6, 50 mg
SDP
Catalog No. 502890874
Encompass_Preferred
An iodinated nonivamide; a TRPV1 antagonist (IC50 = ~10 nM)
Catalog No. 50-289-0874 Supplier Cayman Chemical Supplier No. 1000563350 MG
An iodinated nonivamide; a TRPV1 antagonist (IC50 = ~10 nM)