Learn More
Cayman Chemical CAY10448 Fatty Acids and Deriv
SDP
Supplier:  Cayman Chemical 1000563310 MG
Encompass_Preferred
An iodinated nonivamide; a TRPV1 antagonist (IC50 = ~10 nM)
Catalog No. 50-289-1112