Learn More
Cayman Chemical CAY10448, Purity: ≥98%, CAS: 1177195-52-6, 5 mg
SDP
Catalog No. 502888779
Encompass_Preferred
An iodinated nonivamide; a TRPV1 antagonist (IC50 = ~10 nM)
Catalog No. 50-288-8779 Supplier Cayman Chemical Supplier No. 100056335 MG
An iodinated nonivamide; a TRPV1 antagonist (IC50 = ~10 nM)