Learn More
Cayman Chemical CAY10455 Fatty Acids and Deriv
SDP
Supplier:  Cayman Chemical 10005072100 UG
Encompass_Preferred
A fluorescent substrate for AEA transport; cleavage by intracellular esterases gives a bright fluorescence at 530 nm
Catalog No. 50-289-1228