Learn More
Cayman Chemical CAY10455 Fatty Acids and Deriv
SDP
Catalog No. 502891289
Encompass_Preferred
A fluorescent substrate for AEA transport; cleavage by intracellular esterases gives a bright fluorescence at 530 nm
Catalog No. 50-289-1289 Supplier Cayman Chemical Supplier No. 10005072500 UG
A fluorescent substrate for AEA transport; cleavage by intracellular esterases gives a bright fluorescence at 530 nm