Learn More
Cayman Chemical CAY10455, Purity: ≥98%, CAS: 290374-09-3, 1 mg
SDP
Catalog No. 502890273
Encompass_Preferred
A fluorescent substrate for AEA transport; cleavage by intracellular esterases gives a bright fluorescence at 530 nm
Catalog No. 50-289-0273 Supplier Cayman Chemical Supplier No. 100050721 MG
A fluorescent substrate for AEA transport; cleavage by intracellular esterases gives a bright fluorescence at 530 nm