Learn More
Cayman Chemical CAY10462, Purity: ≥98%, CAS: 502656-68-0, 50 mg
SDP
Catalog No. 502890956
Encompass_Preferred
A selective inhibitor of the 20-HETE synthase CYP4A11 (IC50 = 8.8 nM in human renal microsomes); 200-fold less potent as an inhibitor of 1A, 1C, and 3A CYP450 enzymes
Catalog No. 50-289-0956 Supplier Cayman Chemical Supplier No. 1000640050 MG
A selective inhibitor of the 20-HETE synthase CYP4A11 (IC50 = 8.8 nM in human renal microsomes); 200-fold less potent as an inhibitor of 1A, 1C, and 3A CYP450 enzymes