Learn More
Cayman Chemical CAY10462 Fatty Acids and Deriv
SDP
Supplier:  Cayman Chemical 1000640010 MG
Encompass_Preferred
A selective inhibitor of the 20-HETE synthase CYP4A11 (IC50 = 8.8 nM in human renal microsomes); 200-fold less potent as an inhibitor of 1A, 1C, and 3A CYP450 enzymes
Catalog No. 50-288-8154