Learn More
Cayman Chemical CAY10485, Purity: ≥98%, CAS: 615264-62-5, 50 mg
SDP
Catalog No. 502890430
Encompass_Preferred
An ACAT-1 and -2 inhibitor with antioxidant activity; inhibits human ACAT-1 and ACAT-2 (IC50s = 95 and 81 µM, respectively); inhibits cooper-mediated oxidation of LDL by 91% at 2 µM
Catalog No. 50-289-0430 Supplier Cayman Chemical Supplier No. 1000648250 MG
An ACAT-1 and -2 inhibitor with antioxidant activity; inhibits human ACAT-1 and ACAT-2 (IC50s = 95 and 81 µM, respectively); inhibits cooper-mediated oxidation of LDL by 91% at 2 µM