Learn More
Cayman Chemical CAY10485 Fatty Acids and Deriv
SDP
Supplier:  Cayman Chemical 1000648210 MG
Encompass_Preferred
An ACAT-1 and -2 inhibitor with antioxidant activity; inhibits human ACAT-1 and ACAT-2 (IC50s = 95 and 81 µM, respectively); inhibits cooper-mediated oxidation of LDL by 91% at 2 µM
Catalog No. 50-288-8050