Learn More
Cayman Chemical CAY10486, Purity: ≥98%, CAS: 615264-52-3, 25 mg
SDP
Catalog No. 502890040
Encompass_Preferred
An inhibitor of ACAT; inhibits human ACAT-1 and ACAT-2 (IC50s = ~60 µM for both); inhibits copper-mediated oxidation of LDL by about 20% at 3 µM
Catalog No. 50-289-0040 Supplier Cayman Chemical Supplier No. 1000645225 MG
An inhibitor of ACAT; inhibits human ACAT-1 and ACAT-2 (IC50s = ~60 µM for both); inhibits copper-mediated oxidation of LDL by about 20% at 3 µM