Learn More
Cayman Chemical CAY10486 Fatty Acids and Deriv
SDP
Catalog No. 502890426
Encompass_Preferred
An inhibitor of ACAT; inhibits human ACAT-1 and ACAT-2 (IC50s = ~60 µM for both); inhibits copper-mediated oxidation of LDL by about 20% at 3 µM
Catalog No. 50-289-0426 Supplier Cayman Chemical Supplier No. 100064525 MG
An inhibitor of ACAT; inhibits human ACAT-1 and ACAT-2 (IC50s = ~60 µM for both); inhibits copper-mediated oxidation of LDL by about 20% at 3 µM