Learn More
Cayman Chemical CAY10487, Purity: ≥98%, CAS: 778624-05-8, 5 mg
SDP
Catalog No. 502888194
Encompass_Preferred
An antioxidant with anti-atherogenic activity; inhibits formation of fatty streak lesions of the thoracic aorta in high cholesterol-fed rabbits without affecting plasma lipid profiles or significantly inhibiting ACAT-1 or ACAT-2 activity; dietary administration (0.05%) reduces the percent area occupied by the atherosclerotic lesion in rabbits to 16.1% from 53.5% in control rabbits; inhibits copper-mediated oxidation of LDL by ~75% at 2 µM
Catalog No. 50-288-8194 Supplier Cayman Chemical Supplier No. 100064805 MG
An antioxidant with anti-atherogenic activity; inhibits formation of fatty streak lesions of the thoracic aorta in high cholesterol-fed rabbits without affecting plasma lipid profiles or significantly inhibiting ACAT-1 or ACAT-2 activity; dietary administration (0.05%) reduces the percent area occupied by the atherosclerotic lesion in rabbits to 16.1% from 53.5% in control rabbits; inhibits copper-mediated oxidation of LDL by ~75% at 2 µM