Learn More
Cayman Chemical CAY10499, Purity: ≥98%, CAS: 359714-55-9, 10 mg
SDP
Catalog No. 502888629
Encompass_Preferred
A non-selective lipase inhibitor (IC50s = 144, 90, and 14 nM for human recombinant MAGL, HSL, and FAAH, respectively); inhibits ATGL, DAGLa, ABHD6, and CES1 at 5 mM; inhibits the growth of MCF-7, MDA-MB-231, COV318, and OVCAR-3 cancer cells (IC50s = 4.2, 46, 106.7, and 79.8 µM, respectively); reduces FRUC diet- and AIN-93M diet-induced increases in cytosolic lipase activity in rats
Catalog No. 50-288-8629 Supplier Cayman Chemical Supplier No. 1000787510 MG
A non-selective lipase inhibitor (IC50s = 144, 90, and 14 nM for human recombinant MAGL, HSL, and FAAH, respectively); inhibits ATGL, DAGLa, ABHD6, and CES1 at 5 mM; inhibits the growth of MCF-7, MDA-MB-231, COV318, and OVCAR-3 cancer cells (IC50s = 4.2, 46, 106.7, and 79.8 µM, respectively); reduces FRUC diet- and AIN-93M diet-induced increases in cytosolic lipase activity in rats