Learn More
Medchemexpress LLC HY-13530 10mg Medchemexpress, CAY10505 CAS:1218777-13-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1353010MG
Encompass_Preferred
Medchemexpress, HY-13530 10mg CAY10505 CAS:1218777-13-9 CAY10505 is a potent and selective PI3Kγ inhibitor with an IC50 of 30 nM in neurons. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1381