Learn More
Apexbio Technology LLC CAY10505 1218777-13-9 200mg
SDP
Catalog No. 5000089752
Encompass_Preferred
CAY10505 is a small-molecule inhibitor targeting phosphatidylinositol 3-kinase gamma (PI3K ) It is designed to selectively inhibit PI3K thereby modulating cellular processes governed by the PI3K/Akt/mTOR signaling pathway CAY10505 exerts its biological activity primarily through inhibition of PI3K In cultured neuron studies CAY10505 demonstrates inhibition of baicalein-induced Akt phosphorylation with an IC50 value of approximately 30 nM Based on these pharmacological properties CAY10505 holds research potential in neuroscience cardiovascular biology and cancer biology studies
Catalog No. 50-000-89752 Supplier Apexbio Technology LLC Supplier No. B2182200
CAY10505 is a small-molecule inhibitor targeting phosphatidylinositol 3-kinase gamma (PI3K ) It is designed to selectively inhibit PI3K thereby modulating cellular processes governed by the PI3K/Akt/mTOR signaling pathway CAY10505 exerts its biological activity primarily through inhibition of PI3K In cultured neuron studies CAY10505 demonstrates inhibition of baicalein-induced Akt phosphorylation with an IC50 value of approximately 30 nM Based on these pharmacological properties CAY10505 holds research potential in neuroscience cardiovascular biology and cancer biology studies