Learn More
Cayman Chemical CAY10509 Fatty Acids and Deriv
SDP
Supplier:  Cayman Chemical 10009167500 UG
Encompass_Preferred
Derivative of PGF1α containing a thio-fluorobenzene substituent in the lower side chain; binds to the recombinant human FP receptor with an IC50 value of about 30 nM
Catalog No. 50-289-0377