Learn More
Cayman Chemical CAY10509 Fatty Acids and Deriv
SDP
Catalog No. 502891347
Encompass_Preferred
Derivative of PGF1α containing a thio-fluorobenzene substituent in the lower side chain; binds to the recombinant human FP receptor with an IC50 value of about 30 nM
Catalog No. 50-289-1347 Supplier Cayman Chemical Supplier No. 100091671 MG
Derivative of PGF1α containing a thio-fluorobenzene substituent in the lower side chain; binds to the recombinant human FP receptor with an IC50 value of about 30 nM