Learn More
Cayman Chemical CAY10509, Purity: ≥98%, CAS: 1245699-47-1, 5 mg
SDP
Catalog No. 502888577
Encompass_Preferred
Derivative of PGF1α containing a thio-fluorobenzene substituent in the lower side chain; binds to the recombinant human FP receptor with an IC50 value of about 30 nM
Catalog No. 50-288-8577 Supplier Cayman Chemical Supplier No. 100091675 MG
Derivative of PGF1α containing a thio-fluorobenzene substituent in the lower side chain; binds to the recombinant human FP receptor with an IC50 value of about 30 nM