Learn More
Cayman Chemical CAY10562 Fatty Acids and Deriv
SDP
Catalog No. 502890538
Encompass_Preferred
A S-nitrosothiol that acts as an NO donor under acidic conditions; decomposes with a half-life of 6 minutes in 0.1 M phosphate buffer, pH 5.0, at 37°C; relaxes phenylephrine-constricted rat aortic strips 35% and 8% at pH 6.0 and 7.4, respectively
Catalog No. 50-289-0538 Supplier Cayman Chemical Supplier No. 100105021 MG
A S-nitrosothiol that acts as an NO donor under acidic conditions; decomposes with a half-life of 6 minutes in 0.1 M phosphate buffer, pH 5.0, at 37°C; relaxes phenylephrine-constricted rat aortic strips 35% and 8% at pH 6.0 and 7.4, respectively