Learn More
Medchemexpress LLC HY-15823 1mg Medchemexpress, CAY10566 CAS:944808-88-2 Purity:>98%
SDP
Catalog No. 501932757
Encompass_Preferred
Medchemexpress, HY-15823 1mg CAY10566 CAS:944808-88-2 CAY10566 is a potent, orally bioavailable and selective stearoyl-CoA desaturase1 ( SCD1 ) inhibitor with IC 50 s of 4.5 and 26 nM in mouse and human enzymatic assays, respectively.CAY10566 also shows excellent cellular activity in blocking the conversion of saturated long-chain fatty acid-CoAs (LCFA-CoAs) to monounsaturated LCFA-CoAs in HepG2 cells (IC 50 =7.9 nM or 6.8 nM) [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2757 Supplier Medchemexpress LLC Supplier No. HY158231MG
Medchemexpress, HY-15823 1mg CAY10566 CAS:944808-88-2 CAY10566 is a potent, orally bioavailable and selective stearoyl-CoA desaturase1 ( SCD1 ) inhibitor with IC 50 s of 4.5 and 26 nM in mouse and human enzymatic assays, respectively.CAY10566 also shows excellent cellular activity in blocking the conversion of saturated long-chain fatty acid-CoAs (LCFA-CoAs) to monounsaturated LCFA-CoAs in HepG2 cells (IC 50 =7.9 nM or 6.8 nM) [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.