Learn More
Cayman Chemical CAY10566, 944808-88-2, 5 mg
SDP
Supplier:  Cayman Chemical 100125625
Encompass_Preferred
A potent, selective inhibitor of Stearoyl-CoA desaturase 1 that demonstrates IC50 values of 4.5 and 26 nM in mouse and human enzymatic assays, respectively
Catalog No. NC0444987