Learn More
Medchemexpress LLC HY-15823 25mg , CAY10566 CAS:944808-88-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1582325MG
Encompass_Preferred
HY-15823 25mg CAY10566 CAS: 944808-88-2 CAY10566 is a potent, orally bioavailable and selective stearoyl-CoA desaturase1 (SCD1) inhibitor with IC50s of 4.5 and 26 nM in mouse and human enzymatic assays, respectively.CAY10566 also shows excellent cellular activity in blocking the conversion of saturated long-chain fatty acid-CoAs (LCFA-CoAs) to monounsaturated LCFA-CoAs in HepG2 cells (IC50=7.9 nM or 6.8 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9372