Learn More
Abcam CAY10572 (PHA-767491), Cdc7 and Cdk9 kinase inhibitor, 5MG
Catalog No. 502705764
Encompass_Preferred
MW 249.69 Da, Purity >98%. Potent Cdc7 and Cdk9 kinase inhibitor (IC₅₀ values are 10 and 34 nM for Cdc7 in the presence of 1.5 μM ATP and Cdk9 respectively). Induces apoptosis in cancer cell lines. Shows synergistic antitumor effects in vivo with 5-fluorouracil (ab142387).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5764 Supplier Abcam Supplier No. AB1444195MG
Add to Cart
Add to Cart
MW 249.69 Da, Purity >98%. Potent Cdc7 and Cdk9 kinase inhibitor (IC₅₀ values are 10 and 34 nM for Cdc7 in the presence of 1.5 μM ATP and Cdk9 respectively). Induces apoptosis in cancer cell lines. Shows synergistic antitumor effects in vivo with 5-fluorouracil (ab142387).
The product is subject to the following: Abcam Restricted Use Statement