Learn More
Cayman Chemical CAY10573, Purity: ≥98%, CAS: 853652-40-1, 1 mg
SDP
Catalog No. 502890032
Encompass_Preferred
A PPAR agonist that displays potent binding at PPARα, γ, and δ (IC50s = 113, 50, 223 nM, respectively) and potently transactivates PPARα, γ, and δ (EC50s = 8, 70, 500 nM, respectively); demonstrates stronger binding and functional activity for PPARγ than rosiglitazone
Catalog No. 50-289-0032 Supplier Cayman Chemical Supplier No. 100088461 MG
A PPAR agonist that displays potent binding at PPARα, γ, and δ (IC50s = 113, 50, 223 nM, respectively) and potently transactivates PPARα, γ, and δ (EC50s = 8, 70, 500 nM, respectively); demonstrates stronger binding and functional activity for PPARγ than rosiglitazone