Learn More
Abcam CAY10574, Cdk9 kinase inhibitor, 10MG
Catalog No. 502705758
Encompass_Preferred
MW 218.2 g/mol, Purity >95%. Potent, selective Cdk9 kinase inhibitor (IC₅₀ = 350 nM). Moderate ATP-competitive Cdk2-cyclin E inhibitor (IC₅₀ = 20 μM). Induces cell cycle arrest, decreasing S-phase. Shows antiproliferative effects (IC₅₀ values are 33, 49, 64 and 62 μM for MCF7, HOS, G361, and K562 cells respectively).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5758 Supplier Abcam Supplier No. AB14442310MG
MW 218.2 g/mol, Purity >95%. Potent, selective Cdk9 kinase inhibitor (IC₅₀ = 350 nM). Moderate ATP-competitive Cdk2-cyclin E inhibitor (IC₅₀ = 20 μM). Induces cell cycle arrest, decreasing S-phase. Shows antiproliferative effects (IC₅₀ values are 33, 49, 64 and 62 μM for MCF7, HOS, G361, and K562 cells respectively).
The product is subject to the following: Abcam Restricted Use Statement