Learn More
Cayman Chemical CAY10576, 862812-98-4, 1 mg
SDP
Supplier:  Cayman Chemical 100112491
Encompass_Preferred
Molecular Formula C22H19N3O5S2, Purity ≥95%, Formula Weight 469.5, Synonyms: 5-(5,6-dimethoxy-1H-benzimidazol-1-yl)-3-[[2-(methylsulfonyl)phenyl]methoxy]-2-thiophenecarbonitrile; A benzimidazole analog that selectively inhibits IKK-ε with an IC50 value of 40 nM and is essentially inactive at IKK-α and IKK-β.
Catalog No. NC0444197