Learn More
STEMCELL Technologies CAY10585, Size: 10 mg
SDP
Supplier:  STEMCELL Technologies 72432
Encompass
CAY10585 inhibits hypoxia inducible factor 1 (HIF-1) accumulation in a concentration and time-dependent manner and significantly suppresses transcriptional activity of HIF-1 target genes (Lee et al.).DIFFERENTIATION· Inhibits BMP9- and HIF-1-induced osteogenic differentiation in mesenchymal stem cells (Hu et al.).· Rescues cardiomyocyte differentiation in Fgfr1(-/-) mouse embryonic stem cells (Crescini et al.).
Catalog No. 50-197-5922