Learn More
Cayman Chemical CAY10591, Purity: ≥98%, CAS: 839699-72-8, 10 mg
SDP
Catalog No. 502891381
Encompass_Preferred
An activator of SIRT1 that decreases TNF-α levels from 325 pg/ml (control) to 104 and 53 pg/ml at 20 and 60 µM, respectively; exhibits a significant dose-dependent effect on fat mobilization in differentiated adipocytes
Catalog No. 50-289-1381 Supplier Cayman Chemical Supplier No. 1000979710 MG
An activator of SIRT1 that decreases TNF-α levels from 325 pg/ml (control) to 104 and 53 pg/ml at 20 and 60 µM, respectively; exhibits a significant dose-dependent effect on fat mobilization in differentiated adipocytes