Learn More
Cayman Chemical CAY10591, Purity: ≥98%, CAS: 839699-72-8, 25 mg
SDP
Catalog No. 502887798
Encompass_Preferred
An activator of SIRT1 that decreases TNF-α levels from 325 pg/ml (control) to 104 and 53 pg/ml at 20 and 60 µM, respectively; exhibits a significant dose-dependent effect on fat mobilization in differentiated adipocytes
Catalog No. 50-288-7798 Supplier Cayman Chemical Supplier No. 1000979725 MG
An activator of SIRT1 that decreases TNF-α levels from 325 pg/ml (control) to 104 and 53 pg/ml at 20 and 60 µM, respectively; exhibits a significant dose-dependent effect on fat mobilization in differentiated adipocytes