Learn More
TARGETMOL CHEMICALS INC CAY10594 5MG
SDP
Catalog No. 502579230
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CAY10594 is a potent phospholipase D2(PLD2) inhibitor. CAY10594 ameliorates acetaminophen-induced acute liver injury by regulating the phosphorylated-GSK-3(beta)/JNK axis. purity: 98%
Catalog No. 50-257-9230 Supplier TARGETMOL CHEMICALS INC Supplier No. T72805MG
Add to Cart
Add to Cart
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CAY10594 is a potent phospholipase D2(PLD2) inhibitor. CAY10594 ameliorates acetaminophen-induced acute liver injury by regulating the phosphorylated-GSK-3(beta)/JNK axis. purity: 98%